Ward's® Live Cyanobacteria Mixture
Supplier: Avantor
470179-926EA 17.95 USD
470179-926 470179-930
Ward's® Live Cyanobacteria Mixture
Specimens Live Specimens Bacteria - Living Material
Taking their name from the bluish pigment that most of these organisms share, each jar contains a broad sample of several types of cyanobacteria.
  • Observe various genera without having to purchase multiple specimens
  • Allow your students to identify the various organisms present in the jar
Five or more common types of these aquatic, photosynthetic
cyanobacteria are supplied; specimens may vary in genera, species are not specified.
Genera most commonly included are: Anabaena, Gloeotrichia, Nostoc, Oscillatoria, and Scytonema. Species are not guaranteed.