Ammonium Sulfate

Supplier: ALDON CORP SE

AA0325-100G AA0325-500G AA0325-500GR AA0327-500ML AA0326-500ML
470300-252EA 13 USD
470300-252 470300-254 470300-256 470225-728 470225-750
Ammonium Sulfate
Chemicals for Science Education

CAS Number: 7783-20-2
Formula Weight: 132.15
Formula: (NH4)2SO4
Density (g/mL): 1.769
Solubility: Water
Synonyms: Sulfuric Acid Diammonium Salt
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR