Adjustable Boss Head Clamp Holders
Supplier: GSC INTERNATIONAL SE
4-ACH40 4-ACH40-10
470235-748EA 22.2 USD
470235-748 470121-788
Adjustable Boss Head Clamp Holders
Clamps General Purpose Clamps
This adjustable clamp can rotate 360° and then be locked in place.
  • Nickel plated thumb screws
  • Accepts rods up to ⅝" dia.
Looking for a versatile and reliable way to hold your lab equipment in place?
Crafted with precision, this sturdy clamp features nickel-plated thumb screws that allow you to securely grip rods up to ⅝" diameter. But what really sets it apart is its ability to adjust to any angle, giving you complete control and flexibility in your setup.
Whether you're conducting experiments, assembling delicate equipment, or simply need a reliable way to hold your glassware, the clamp holder has got your back. Durable, easy to use, this is a must-have tool for any science, lab, or STEAM-focused environment.
Stop struggling with awkward setups and invest in the clamp holder from boss head - the straightforward solution for all your laboratory needs.