Acetanilide

Supplier: ALDON CORP SE

AA0018-100G
470300-026EA 92.15 USD
470300-026
Acetanilide
Chemicals for Science Education

CAS Number: 103-84-4
Formula Weight: 135.17
Formula: CH3CONHC6H5
Density (g/mL): 1.2
Boiling Point (°C): 304
Freezing Point (°C): 115
Solubility: Hot Water, Alcohol, and Organic Solvents
Synonyms: N-phenyl-acetamide
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR