(+)-Valencene
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Valencene (Standard), (3R,4aS,5R)-4a,5-Dimethyl-3-(prop-1-en-2-yl)-1,2,3,4,4a,5,6,7-octahydronaphthalene
V1013-100GM
V1013-500GM
V1013-25GM
700002-404EA
111.68
USD
700002-404
700002-398
700002-402
700002-400
(+)-Valencene
(+)-Valencene
(+)-Valencene is a sesquiterpeneone of the main contributors to the flavor and odor of citrus.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₅H₂₄ MW: 204.36 g/mol Density: 0.92 Storage Temperature: Refrigerator |
MDL Number:
MFCD00075884 CAS Number: 4630-07-3 EINECS: 225-047-6 |
Specification Test Results
Assay | 65% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...