Potassium bromide ≥99%, crystals ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
P1220-25GM
P1220-25KGBL
P1220-12KG
P1220-500GM
P1220-125GM
700001-246EA
89.31
USD
700001-246
700001-248
700001-242
700001-250
700001-240
Potassium bromide ≥99%, crystals ACS
Potassium bromide
Potassium Bromide, Crystal, Reagent, ACS, is found as a white crystal and is very soluble in water. In infrared spectroscopy, samples are analyzed by grinding with powdered potassium bromide and pressing into a disc. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 °C (1013 hPa) Melting Pt: 730 °C Density: 2.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 13,07701 |
Specification Test Results
Assay | 99% Min. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...