Cysteaminium chloride ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

C2722-100GM C2722-25GM C2722-50KGBL
700008-034EA 284.95 USD
700008-034 700008-036 700008-038
Cysteaminium chloride ≥98%
Cysteaminium chloride

Cysteamine Hydrochloride is a stable aminothiol and a product of the amino acid cysteine. It has been used to treat radiation sickness.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂H₇NS·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR