Blue Dextran loading dye

Supplier: G-Biosciences

Synonyms: Dextran, blue

786-574 786-556 786-557
71003-188EA 200.27 USD
71003-188 89167-764 89167-766
Blue Dextran loading dye
Blue Dextran

A proprietary mixture of Formaldehyde, EDTA and Blue Dextran that is designed for use in loading DNA samples onto denaturing polyacrylamide gels.


Commonly used in loading DNA samples onto denaturing polyacrylamide gels.

Formula: (C₂₉H₂₁N₇O₁₂S₃·3Na)n
MDL Number: MFCD00130661
CAS Number: 87915-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR