2-Chloro-3-fluoropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20022-03
L20022-06
AAAL20022-03EA
45.1
USD
AAAL20022-03
AAAL20022-06
2-Chloro-3-fluoropyridine 97%
2-Chloro-3-fluoropyridine
Purity: 97%
CAS No.: 17282-04-1
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Flash Point: 72°(161°F)
Refractive Index: 1.5095
Odor: Unpleasant
Boiling Point: 62-64°/23mm
Beilstein Reference: 1363435
UN No.: 2810
MDL No.: MFCD03095302
Formula:
C₅H₃ClFN Boiling Pt: 62…64 °C/23 mm Density: 1.325 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03095302 CAS Number: 17282-04-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...