Trioctylphosphine tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TOP, Tri-n-octylphosphine

L15744-AC L15744-AE L15744-AP
AAAL15744-ACEA 58.05 USD
AAAL15744-AC AAAL15744-AE AAL15744-AP
Trioctylphosphine tech. 90%, Technical Grade
Trioctylphosphine

Purity: 90% CAS No.: 4731-53-7 Molecular Formula: C24H51P Formula Weight: 370.65 Flash Point: 147°(296°F) Refractive Index: 1.4680 Odor: Garlic-like Boiling Point: 240°/10mm Beilstein Reference: 1776995 UN No.: 1760 MDL No.: MFCD00015298

Formula: C₂₄H₅₁P
Boiling Pt: 240 °C/10 mm
Melting Pt: 48 °C
Density: 0.831
Flash Pt: 147 °C(296 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015298
CAS Number: 4731-53-7
EINECS: 225-234-2
UN: 1760
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR