4-Iodophenylhydrazine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(4-Iodophenyl)hydrazine
L14806-06
L14806-03
AAAL14806-06EA
279
USD
AAAL14806-06
AAAL14806-03
4-Iodophenylhydrazine 95%
4-Iodophenylhydrazine
Purity: 95%
CAS No.: 13116-27-3
Molecular Formula: C6H7IN2
Formula Weight: 234.04
Storage Temperature: 2°C to 8°C
Melting Point: 99-103°
Beilstein Reference: 742475
MDL No.: MFCD00052688
Formula:
C₆H₇IN₂ MW: 234.04 g/mol Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052688 CAS Number: 13116-27-3 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...