Triethylammonium dihydrogentrifluoride (∼37% HF)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Triethylamine trihydrofluoride, Triethylamine trishydrofluoride
L14417-06
L14417-14
L14417-22
AAAL14417-06EA
29.12
USD
AAAL14417-06
AAAL14417-14
AAAL14417-22
Triethylammonium dihydrogentrifluoride (∼37% HF)
Triethylammonium dihydrogentrifluoride
CAS No.: 73602-61-6
Molecular Formula: C6H15N.3HF
Formula Weight: 161.21
Flash Point: 87°(188°F)
Refractive Index: 1.3915
Melting Point: -29°
Boiling Point: 70°/15mm
Beilstein Reference: 5522945
UN No.: 1790
MDL No.: MFCD00043294
Formula:
C₆H₁₈F₃N MW: 161.21 Boiling Pt: 70 °C/15 mm Melting Pt: –29 °C Density: 0.990 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043294 CAS Number: 73602-61-6 EINECS: 277-550-5 UN: 1790 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...