Magnesium ≥99%, turnings
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08120-36
L08120-0E
L08120-22
AAAL08120-36EA
90.6
USD
AAAL08120-36
AAL08120-0E
AAL08120-22
Magnesium ≥99%, turnings
Magnesium
Purity: 99+%
CAS No.: 7439-95-4
Odor: Odorless
UN No.: 1869
MDL No.: MFCD00085308
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Density: 1.738 Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1 III Merck Index: 13,05675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...