Acetic anhydride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetyl oxide, Acetyl acetate
L04295-0B
L04295-30
L04295-0E
AAAL04295-0BEA
103.44
USD
AAAL04295-0B
AAAL04295-30
AAAL04295-0E
Acetic anhydride 99+%
Acetic anhydride
Purity: 99+%
CAS No.: 108-24-7
Molecular Formula: (CH3CO)2O
Formula Weight: 102.09
Flash Point: 49°(120°F)
Refractive Index: 1.3900
Odor: Pungent
Melting Point: -73°
Boiling Point: 138-140°
Beilstein Reference: 385737
Merck Reference: 14,56
UN No.: 1715
MDL No.: MFCD00008705
Formula:
(CH₃CO)₂O MW: 102.09 g/mol Boiling Pt: 138…140 °C Melting Pt: –73 °C Density: 1.082 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008705 CAS Number: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II REACH: 01-2119486470-36 Merck Index: 14,56 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...