Triphenylarsine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03616-06
L03616-14
AAAL03616-06EA
100
USD
AAAL03616-06
AAAL03616-14
Triphenylarsine 97%
Triphenylarsine
Purity: 97%
CAS No.: 603-32-7
Molecular Formula: C18H15As
Formula Weight: 306.24
Flash Point: 265°(509°F)
Melting Point: 58-62°
Boiling Point: 360°
Beilstein Reference: 1842037
UN No.: 3465
MDL No.: MFCD00002994
Formula:
C₁₈H₁₅As MW: 306.24 g/mol Boiling Pt: 360 °C Melting Pt: 58 Flash Pt: 265 °C (509 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002994 CAS Number: 603-32-7 EINECS: 210-032-9 UN: 3465 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...