1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 1,2-Diphenyltetramethyldisilane

H28705-03 H28705-06
AAH28705-03EA 67.5 USD
AAH28705-03 AAH28705-06
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
1,2-Diphenyl-1,1,2,2-tetramethyldisilane

Purity: 95% CAS No.: 1145-98-8 Molecular Formula: C16H22Si2 Formula Weight: 270.52 MDL No.: MFCD00054786

Formula: C₁₆H₂₂Si₂
MW: 270.52 g/mol
Boiling Pt: 73
Melting Pt: 37
Storage Temperature: Ambient
MDL Number: MFCD00054786
CAS Number: 1145-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR