2-Chlorolepidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-4-methylquinoline

H26504-06 H26504-14
AAH26504-06EA 52.2 USD
AAH26504-06 AAH26504-14
2-Chlorolepidine 99%
2-Chlorolepidine

Purity: 99% CAS No.: 634-47-9 Molecular Formula: C10H8ClN Formula Weight: 177.63 Melting Point: 54-56° Boiling Point: 296° MDL No.: MFCD00006742

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Boiling Pt: 296
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00006742
CAS Number: 634-47-9
EINECS: 211-209-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR