3-Fluoro-4-nitrobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

H26168-03 H26168-06
AAH26168-03EA 20.2 USD
AAH26168-03 AAH26168-06
3-Fluoro-4-nitrobenzoic acid 98+%
3-Fluoro-4-nitrobenzoic acid

Purity: 98+% CAS No.: 403-21-4 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Melting Point: 173-176° MDL No.: MFCD01862092

Formula: C₇H₄FNO₄
MW: 185.11
Melting Pt: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD01862092
CAS Number: 403-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR