4-Ethylbenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Ethylbenzene-1-sulfonyl chloride, 4-Ethylbenzene-1-sulphonyl chloride
B24682-06
B24682-14
AAAB24682-22EA
621.43
USD
AAAB24682-22
AAAB24682-06
AAAB24682-14
4-Ethylbenzenesulfonyl chloride 97%
4-Ethylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 16712-69-9
Molecular Formula: C8H9ClO2S
Formula Weight: 204.68
Refractive Index: 1.5450
Melting Point: 8-12°
Boiling Point: 134-136°/7mm
Beilstein Reference: 2090130
UN No.: 3265
MDL No.: MFCD00041480
Formula:
C₈H₉ClO₂S Boiling Pt: 134…136 °C/7 mm Melting Pt: 8…12 °C Density: 1.268 Storage Temperature: Ambient |
MDL Number:
MFCD00041480 CAS Number: 16712-69-9 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...