5-Bromo-2-iodotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning

B21878-22 B21878-14
AAAB21878-06EA 34.95 USD
AAAB21878-06 AAAB21878-22 AAB21878-14
5-Bromo-2-iodotoluene 98+%
5-Bromo-2-iodotoluene

Purity: 98+% CAS No.: 116632-39-4 Molecular Formula: C7H6BrI Formula Weight: 296.93 Flash Point: 110°(230°F) Refractive Index: 1.6515 Boiling Point: 262-265° Beilstein Reference: 2325157 MDL No.: MFCD00060664

Formula: C₇H₆BrI
Boiling Pt: 262…265 °C
Density: 2.139
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060664
CAS Number: 116632-39-4
EINECS: 629-570-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR