Sorbic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

A16196-0B A16196-0I A16196-30 A16196-36
AAAA16196-0BEA 112.19 USD
AAAA16196-0B AAAA16196-0I AAAA16196-30 AAAA16196-36
Sorbic acid 99%
Sorbic acid

Purity: 99% CAS No.: 110-44-1 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 127°(260°F) Melting Point: 132-135° Boiling Point: 228° dec. Beilstein Reference: 1741831 Merck Reference: 14,8721 MDL No.: MFCD00002703

Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228 (dec.)
Melting Pt: 132…135 °C
Density: 1.205
Flash Pt: 127 °C (260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 14,8721
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR