Triphosgene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(trichloromethyl) carbonate
A14932-14
A14932-22
A14932-06
AAAA14932-14EA
179
USD
AAAA14932-14
AAAA14932-22
AAAA14932-06
Triphosgene 98%
Triphosgene
Purity: 98%
CAS No.: 32315-10-9
Molecular Formula: C3Cl6O3
Formula Weight: 296.75
Storage Temperature: 2°C to 8°C
Melting Point: 77-91°
Boiling Point: 203-206°
Beilstein Reference: 1787583
UN No.: 2928
MDL No.: MFCD00062848
Formula:
C₃Cl₆O₃ MW: 296.75 g/mol Boiling Pt: 203…206 °C Melting Pt: 77…91 °C Density: 1.600 Storage Temperature: Refrigerator |
MDL Number:
MFCD00062848 CAS Number: 32315-10-9 EINECS: 250-986-3 UN: 2928 ADR: "6.1",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...