Ethylenetetracarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetracyanoethylene

A13945-03 A13945-06
AAAA13945-03EA 33.79 USD
AAAA13945-03 AAAA13945-06
Ethylenetetracarbonitrile 98%
Ethylenetetracarbonitrile

Purity: 98% CAS No.: 670-54-2 Molecular Formula: C6N4 Formula Weight: 128.09 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 1.348 Sensitivity: Moisture Sensitive Melting Point: 197-199° Boiling Point: 223° Beilstein Reference: 1679885 Merck Reference: 14,9195 UN No.: 3439 MDL No.: MFCD00001850

Formula: C₆N₄
MW: 128.09 g/mol
Boiling Pt: 223 °C
Melting Pt: 197…199 °C
Density: 1.348
Storage Temperature: Refrigerator
MDL Number: MFCD00001850
CAS Number: 670-54-2
EINECS: 211-578-0
UN: 3439
ADR: "6.1",III
Merck Index: 14,9195

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR