Triiodomethane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Iodoform

A13664-0B A13664-30 A13664-18
AAAA13664-0BEA 529 USD
AAAA13664-0B AAAA13664-30 AAAA13664-18
Triiodomethane 99%
Triiodomethane

Purity: 99% CAS No.: 75-47-8 Molecular Formula: CHI3 Formula Weight: 393.73 Flash Point: None Odor: Unpleasant Beilstein Reference: 1697010 Merck Reference: 14,5033 MDL No.: MFCD00001069

Formula: CHI₃
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 118…123 °C
Density: 4.008
Storage Temperature: Ambient
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5
Merck Index: 14,5033

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR