Potassium permanganate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Permanganic acid potassium salt

A12170-0E A12170-36 A12170-0C
AAAA12170-0EEA 168 USD
AAAA12170-0E AAAA12170-36 AAAA12170-0C
Potassium permanganate 98%
Potassium permanganate

Purity: 98% CAS No.: 7722-64-7 Molecular Formula: KMnO4 Formula Weight: 158.03 Melting Point: 240° dec. Merck Reference: 14,7655 MDL No.: MFCD00011364

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: 240 °C (dec.)
Density: 2.7
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 14,7655

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR