Pyrocatechol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 120-80-9
Molecular Formula: C6H6O2
Formula Weight: 110.11
Flash Point: 127°(260°F)
Physical Form: Powder or flakes
Density: 1.370
Sensitivity: Air and Light Sensitive
Appearance: White to brown
Odor: Aromatic
Melting Point: 104-107°
Boiling Point: 245-246°
Beilstein Reference: 471401
Merck Reference: 14,7999
UN No.: 2811
MDL No.: MFCD00002188
Formula:
C₆H₆O₂ Boiling Pt: 245…246 °C Melting Pt: 104…107 °C Density: 1.370 Flash Pt: 127 °C (260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002188 CAS Number: 120-80-9 EINECS: 204-427-5 UN: 2811 ADR: 6.1,III Merck Index: 14,7999 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...