Copper, gauze woven from wire 100 mesh, Diameter 0.11 mm (0.0045 in)

Supplier: Thermo Scientific Chemicals

045186-GT 045186-HB
AA45186-GTEA 31.4 USD
AA45186-GT AA45186-HB
Copper, gauze woven from wire 100 mesh, Diameter 0.11 mm (0.0045 in)
Copper

CAS No.: 7440-50-8 Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR