Copper, gauze woven from wire 60 mesh, Diameter 0.19 mm (0.0075 in)

Supplier: Thermo Scientific Chemicals

045142-GT 045142-HB
AA45142-GTEA 33.1 USD
AA45142-GT AA45142-HB
Copper, gauze woven from wire 60 mesh, Diameter 0.19 mm (0.0075 in)
Copper

CAS No.: 7440-50-8 Physical Form: Wire cloth Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR