Gold ≥99.999% (metals basis), wire, Premion®, Diameter 0.35 mm (0.0138 in)

Supplier: Thermo Scientific Chemicals

045084-BY 045084-CC 045084-BU
AA45084-BYEA 551 USD
AA45084-BY AA45084-CC AA45084-BU
Gold ≥99.999% (metals basis), wire, Premion®, Diameter 0.35 mm (0.0138 in)
Gold

Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈1.82g/m Odor: Odorless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR