Palladium ≥99.95% (metals basis), wire, hard, Diameter 0.203 mm

Supplier: Thermo Scientific Chemicals

045072-G1 045072-G6
AA45072-G1EA 62.1 USD
AA45072-G1 AA45072-G5 AA45072-G6
Palladium ≥99.95% (metals basis), wire, hard, Diameter 0.203 mm
Palladium

Purity: 99.95% CAS No.: 7440-05-3 Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR