Lead ≥99.8% (metals basis), foil, Thickness 0.76 mm (0.03 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044961-RF
044961-VA
AA44961-RFEA
81.4
USD
AA44961-RF
AA44961-VA
Lead ≥99.8% (metals basis), foil, Thickness 0.76 mm (0.03 in)
Lead
Purity: 99.8%
CAS No.: 7439-92-1
Physical Form: ≈208g/15x15cm
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00134050
Formula:
Pb MW: 207.2 g/mol Boiling Pt: 1740 °C (1013 hPa) Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...