Iron(III) oxide, powder, NanoArc® 20-40nm APS, S.A. 30-60m2/g for cosmetic
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
NanoArc® cosmetic iron oxide is an auburn material offering unique cosmetic qualitites. The small particle size results in a strong color development combined with high translucency for a unique glowing look.
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: 20-40nm APS Powder, S.A. 30-60m2/g
Density: 5.24
Appearance: Red-brown
Odor: Odorless
Melting Point: 1565°
MDL No.: MFCD00011008
NanoArc® cosmetic iron oxide is an auburn material offering unique cosmetic qualitites. The small particle size results in a strong color development combined with high translucency for a unique glowing look
Formula:
Fe₂O₃ Melting Pt: 1565 °C Density: 5.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 13,04058 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...