Gold ≥99.998% (metals basis), powder, Premion® -100 mesh

Supplier: Thermo Scientific Chemicals

044882-03 044882-06
AA44882-03EA 294.33 USD
AA44882-03 AA44882-06
Gold ≥99.998% (metals basis), powder, Premion® -100 mesh
Gold

Purity: 99.998% CAS No.: 7440-57-5 MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR