Chromium ≥99.996% (metals basis), chunks 2-15 mm (0.08-0.59 in)
Supplier: Thermo Scientific Chemicals
044565-09
044565-18
AA44565-09EA
109
USD
AA44565-09
AA44565-18
Chromium ≥99.996% (metals basis), chunks 2-15 mm (0.08-0.59 in)
Chromium
Purity: 99.996%
CAS No.: 7440-47-3
Odor: Odorless
MDL No.: MFCD00010944
Formula:
Cr Boiling Pt: 2469 °C (1013 hPa) Melting Pt: 1840 °C Density: 7.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 Merck Index: 12,02288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...