Copper ≥99.9999% (metals basis), wire, Puratronic®, Diameter 1.5 mm (0.06 in)
Supplier: Thermo Scientific Chemicals
044551-G5
044551-G1
AA44551-G5EA
541
USD
AA44551-G5
AA44551-G1
Copper ≥99.9999% (metals basis), wire, Puratronic®, Diameter 1.5 mm (0.06 in)
Copper
Purity: 99.9999%
CAS No.: 7440-50-8
Physical Form: ≈15.8g/m
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...