Sodium chlorate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

044408-22 044408-36 044408-A3
AA44408-22EA 45.35 USD
AA44408-22 AA44408-36 AA44408-A3
Sodium chlorate ≥99.0% ACS
Sodium chlorate

Purity: 99.0% min CAS No.: 7775-09-9 Molecular Formula: NaClO3 Formula Weight: 106.45 Physical Form: Crystalline Density: 2.5 Melting Point: 248° Merck Reference: 14,8598 UN No.: 1495 MDL No.: MFCD00003479

Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300
Melting Pt: 260
Density: 2.5
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1 II
REACH: 01-2119474389-23
Merck Index: 14,8598

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR