Iron ≥99.9% (metals basis), sponge -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

043240-22 043240-36 043240-A3
AA43240-22EA 34.5 USD
AA43240-22 AA43240-36 AA43240-A3
Iron ≥99.9% (metals basis), sponge -100 mesh
Iron

Purity: 99.9% CAS No.: 7439-89-6 Appearance: Grey UN No.: 3089 MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1 III
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR