Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
010670-03
010670-09
010670-18
AAA10670-03EA
49.6
USD
AAA10670-03
AAA10670-09
AAA10670-18
Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®
Phosphorus
Purity: 99.999%
CAS No.: 7723-14-0
Physical Form: Solid
UN No.: 1338
MDL No.: MFCD00133771
Formula:
P MW: 30.97 g/mol Density: ∼2.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133771 CAS Number: 7723-14-0 EINECS: 231-768-7 UN: 1338 ADR: 4.1,III |
Specification Test Results
Form | Reddish-brown solid |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...