Nalgene® Narrow-Mouth Boston Round Bottles, Natural PPCO, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312006-0001 312006-0002 312006-0004 312006-0008 312006-0016 312006-0032
16059-806CS 1560.96 USD
16059-806 16059-808 16059-810 16059-812 16059-814 16059-816
Nalgene® Narrow-Mouth Boston Round Bottles, Natural PPCO, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
These polypropylene copolymer narrow-mouth, Boston round bottles are feature polypropylene closures.
  • Durable
  • Translucent
  • Autoclavable
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.