Potassium chloride, crystal rectangle for spectrophotometry, polished
Supplier: INTERNATIONAL CRYSTAL LABS MS
Synonyms:
Potassiumchloride
0002B-156
0002B-268
0002B-352
100213-326EA
75.57
USD
100213-326
100211-856
100215-844
Potassium chloride, crystal rectangle for spectrophotometry, polished
Potassium chloride
Potassium Chloride (KCl) is commonly used for infrared transmission crystal windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers. KCl crystal windows are particularly useful for spatter barrier windows in CO2 lasers becaus
Formula:
KCl MW: 74.55 g/mol Boiling Pt: 1420 °C (1013 hPa) Melting Pt: 773 °C Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...