Zinc sulfide, crystal rectangle for spectrophotometry, polished
Supplier: INTERNATIONAL CRYSTAL LABS MS
Synonyms:
Zinc (II) sulphide, Zinc (II) sulfide
0002I-528
0002I-116
0002I-117
0002I-529
100215-424EA
847.69
USD
100215-424
100213-312
100213-314
100216-090
Zinc sulfide, crystal rectangle for spectrophotometry, polished
Zinc sulphide
Zinc Sulfide (ZnS) is used as a transmission window for IR spectroscopy. Zinc Sulfide (ZnS) is or has been produced under the trademarks Cleartran® and Irtran® .
Formula:
SZn MW: 97.46 g/mol Melting Pt: 1064 °C Density: 4.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 12,10294 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...