Zirconium ≥99.5%, powder -50 Mesh for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Z1047-500GM
Z1047-100GM
Z1047-25GM
700013-306EA
1316.05
USD
700013-306
700013-302
700013-304
Zirconium ≥99.5%, powder -50 Mesh for synthesis
Zirconium
Zirconium, -50 Mesh Powder is a lustrous, grey-white, strong transition metal used in small amounts as an alloying agent for its strong resistance to corrosion in environments that are exposed to aggressive environments, such as surgical appliances, light filaments and watch cases.
Formula:
Zr Boiling Pt: 3850 °C (1013 hPa) Melting Pt: 1852 °C Density: 6.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 UN: 1358 ADR: 4.1,II Merck Index: 12,10306 |
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...