Tetradecyltrimethylammonium bromide ≥99%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

T1027-50KGBL T1027-12KGBL T1027-500GM
700007-646EA 3073.05 USD
700007-646 700007-640 700007-644
Tetradecyltrimethylammonium bromide ≥99%, Reagent Grade for synthesis
Tetradecyltrimethylammonium bromide

Tetradecyltrimethylammonium Bromide, Reagent

Formula: C₁₇H₃₈BrN
MW: 336.394 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011770
CAS Number: 1119-97-7
EINECS: 214-291-9
UN: 1759
ADR: 8 II
Merck Index: 12,06419

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR