Sudan IV
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol
Sudan IV is a lysochrome (fat-soluble dye) diazo dye used for the staining of lipids, triglycerides and lipoproteins on frozen paraffin sections. It has the appearance of reddish brown crystals with melting point 199 °C and maximum absorption at 520(357) nm
Formula:
C₂₄H₂₀N₄O MW: 380.45 g/mol Melting Pt: 188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003893 CAS Number: 85-83-6 EINECS: 201-635-8 Merck Index: 12,08538 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...