Squalene ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene

S2002-100ML S2002-500MLG S2002-200LTB S2002-5GLBL
700002-312EA 94.55 USD
700002-312 700002-316 700002-314 700002-318
Squalene ≥99%
Squalene

Squalene, is an organic compound that be found occurring naturally in shark liver oil and certain plants. It is a hydrocarbon that all plants and animals produce in varying quantities. Mostly used in cosmetics as a natural skin moisturizer, it has recently been used as an immunologic adjuvant in certain vaccines

Formula: C₃₀H₅₀
MW: 410.73 g/mol
Boiling Pt: 429 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.858
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00008912
CAS Number: 111-02-4
EINECS: 203-826-1
Merck Index: 13,08847

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR