Sodium chlorite, anhydrous, soluble for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1243-2.5KG S1243-45KGBL S1243-500GM
700001-328EA 1101.15 USD
700001-328 700001-326 700001-330 700001-332
Sodium chlorite, anhydrous, soluble for synthesis
Sodium chlorite

Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis.

Formula: ClNaO₂
MW: 90.44 g/mol
Boiling Pt: 112
Melting Pt: 250…257 °C
Density: 1.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1 II
Merck Index: 13,08672

Order Now

Specification Test Results

Assay 77.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR