D-(+)-Raffinose pentahydrate ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Gossypose pentahydrate
, Melitose pentahydrate
, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside
, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate
, D-(+)-Raffinose pentahydrate
, Melitriose pentahydrate
Raffinose, Pentahydrate, is found naturally in vegetables and whole grains. It is a trisaccharide made up from galactose, glucose and fructose. Raffinose can be used as a base substance for sucralose. It is also used during cryo-preservation procedures to provide hypertonicity for cell desiccation before freezing.
Formula:
C₁₈H₃₂O₁₆·5H₂O MW: 594.52 g/mol Melting Pt: 80 °C Density: 1.516 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071590 CAS Number: 17629-30-0 EINECS: 208-146-9 Merck Index: 12,08279 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...