Piperazine, anhydrous ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

PI123-5GM PI123-100GM PI123-500GM
700008-830EA 98.15 USD
700008-830 700008-820 700008-826
Piperazine, anhydrous ≥99%
Piperazine

Piperazine, Anhydrous is an organic compound that comes as a small deliquescent crystal, is salty to taste and is alkaline. It is used as an anthelminitic to paralyze parasites and in the manufacture of plastics, brake fluid as well as resins and other industrial materials.

Formula: C₄H₁₀N₂
MW: 86.14 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: 44…46 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 13,07545

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR