(±)-1,2-Propanediol ≥99.5% ACS for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
1,2-Propanediol, Reagent, ACS or propylene glycol is an organic compound and colorless liquid without odor. It has a slightly sweet taste and chemically classed as a diol. It is considered generally recognized as safe (GRAS) by the U.S. Food and Drug Administration, and it is used as an humectant (E1520), solvent, and preservative in food and for tobacco products, as well as being one of the major ingredients of the "e-liquid" used in electronic cigarettes along with vegetable glycerin.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
CH₃CH(OH)CH₂OH MW: 76.1 g/mol Boiling Pt: 187.6 Melting Pt: -59 Density: 1.04 g/cm³ (20 °C) Flash Pt: 99 Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 EINECS: 200-338-0 Merck Index: 13,07947 |
|
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...