Potassium peroxodisulfate ≥99%, crystals, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Danger

P1375-125GM P1375-2.5KG P1375-12KG P1375-500GM
700011-842EA 81.95 USD
700011-842 700011-846 700011-844 700011-848
Potassium peroxodisulfate ≥99%, crystals, Reagent Grade
Potassium peroxodisulphate

Potassium Persulfate, Crystal, Reagent, also known as potassium peroxydisulfate, is a strong oxidant used to initiate polymerizations. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: K₂S₂O₈
Melting Pt: 100 °C
Density: 2.477 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 13,07741

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR